ENA150999186 Enamine

5-(1-chloroethyl)-3-(propan-2-yl)-1,2,4-oxadiazole

  • MDL number MFCD09802198
  • Cas Number : 119738-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-29696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29696
Availability details for
Quantity
  • Availability
  • Add to Cart