ENA150999187 Enamine

5-(1-chloroethyl)-3-propyl-1,2,4-oxadiazole

  • MDL number MFCD09802199
  • Cas Number : 119738-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29697

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29697
Availability details for
Quantity
  • Availability
  • Add to Cart