ENA150999190 Enamine

5-(chloromethyl)-1-cyclopropyl-1H-1,2,3,4-tetrazole

  • MDL number MFCD09802201
  • Cas Number : 949980-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29702
Availability details for
Quantity
  • Availability
  • Add to Cart