ENA150999206 Enamine

2-chloro-N-propylpropanamide

  • MDL number MFCD09802012
  • Cas Number : 94318-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23354
Availability details for
Quantity
  • Availability
  • Add to Cart