ENA150999217 Enamine

2-[(piperazin-1-yl)methyl]quinazolin-4-amine

  • MDL number MFCD09802018
  • Cas Number : 949977-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24058
Availability details for
Quantity
  • Availability
  • Add to Cart