ENA150999221 Enamine

2-cyano-3-{2,5-dimethyl-1-[3-(trifluoromethyl)phenyl]-1H-pyrrol-3-yl}acrylic acid

  • MDL number MFCD04638791
  • Cas Number : 736949-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24386

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24386
Availability details for
Quantity
  • Availability
  • Add to Cart