ENA150999238 Enamine

2-(4-Amino-phenyl)-1,1,1,3,3,3-hexafluoro-propan-2-ol

  • MDL number MFCD00039258
  • Cas Number : 722-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02317

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-02317
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart