ENA150999244 Enamine

3-chloro-N-(naphthalen-1-yl)propanamide

  • MDL number MFCD02812237
  • Cas Number : 22302-59-6
  • Molecular Weight : 233.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25817
Availability details for
Quantity
  • Availability
  • Add to Cart