ENA150999245 Enamine

N-[3-(aminosulfonyl)phenyl]-3-chloropropanamide

  • MDL number MFCD09802031
  • Cas Number : 949984-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25818

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25818
Availability details for
Quantity
  • Availability
  • Add to Cart