ENA150999246 Enamine

3-chloro-N-(5-chloro-2-methoxyphenyl)propanamide

  • MDL number MFCD01648150
  • Cas Number : 346726-62-3
  • Molecular Weight : 248.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25820
Availability details for
Quantity
  • Availability
  • Add to Cart