ENA152120876 Enamine

4-(4-fluorophenyl)-5-nitro-1H-imidazole

  • MDL number MFCD09863307
  • Cas Number : 209736-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29857
Availability details for
Quantity
  • Availability
  • Add to Cart