ENA152120877 Enamine

4-chloro-2-(5-chloro-2,3-dihydro-1,3-benzothiazol-2-ylidene)-3-oxobutanenitrile

  • Cas Number : 1000931-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29883
Availability details for
Quantity
  • Availability
  • Add to Cart