ENA152120883 Enamine

5-[(2-chloro-3-methylphenyl)methyl]-2,3-dihydro-1,3-thiazol-2-imine

  • MDL number MFCD21091623
  • Cas Number : 1000931-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29923
Availability details for
Quantity
  • Availability
  • Add to Cart