ENA152120885 Enamine

2-[(propan-2-yloxy)methyl]benzonitrile

  • MDL number MFCD09863327
  • Cas Number : 1000931-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29947
Availability details for
Quantity
  • Availability
  • Add to Cart