ENA152120887 Enamine

(1-amino-3-methylbutan-2-yl)bis(prop-2-en-1-yl)amine

  • MDL number MFCD09863329
  • Cas Number : 1000931-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29949
Availability details for
Quantity
  • Availability
  • Add to Cart