ENA152120891 Enamine

1-methyl-4-sulfamoyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD09863334
  • Cas Number : 1000931-97-4
  • Molecular Weight : 203.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-30408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30408
Availability details for
Quantity
  • Availability
  • Add to Cart