ENA152120896 Enamine

N-cyclopropyl-4-[(cyclopropylamino)methyl]benzamide

  • MDL number MFCD09863339
  • Cas Number : 1000932-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30436

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-30436
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart