ENA152120901 Enamine

1-methyl-2-oxopiperidine-4-carboxylic acid

  • MDL number MFCD09863345
  • Cas Number : 1000932-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30458
Availability details for
Quantity
  • Availability
  • Add to Cart