ENA152120904 Enamine

1-(3-methoxybenzoyl)piperazine hydrochloride

  • MDL number MFCD09863349
  • Cas Number : 100939-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30479
Availability details for
Quantity
  • Availability
  • Add to Cart