ENA152120908 Enamine

4,5-dibromo-1-methyl-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09863354
  • Cas Number : 66067-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30497
Availability details for
Quantity
  • Availability
  • Add to Cart