ENA152120913 Enamine

4-bromo-N-(4-hydroxyphenyl)-N-methylbenzene-1-sulfonamide

  • MDL number MFCD00588500
  • Cas Number : 1000932-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30516
Availability details for
Quantity
  • Availability
  • Add to Cart