ENA152120916 Enamine

4-chloro-2-(1-methyl-1,4-dihydropyridin-4-ylidene)-3-oxobutanenitrile

  • MDL number MFCD09863361
  • Cas Number : 1000932-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30529
Availability details for
Quantity
  • Availability
  • Add to Cart