ENA152120921 Enamine

1-(naphthalene-1-carbonyl)piperazine hydrochloride

  • MDL number MFCD09863366
  • Cas Number : 1093390-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30543
Availability details for
Quantity
  • Availability
  • Add to Cart