ENA152120926 Enamine

1-{[5-(2-fluorophenyl)thiophen-2-yl]methyl}piperazine

  • MDL number MFCD09863371
  • Cas Number : 1000932-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30582

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30582
Availability details for
Quantity
  • Availability
  • Add to Cart