ENA152120943 Enamine

2-nitro-4-trifluoromethanesulfonylbenzene-1-sulfonyl chloride

  • MDL number MFCD09863388
  • Cas Number : 1000932-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30657
Availability details for
Quantity
  • Availability
  • Add to Cart