ENA152120955 Enamine

3-(2-bromoethyl)imidazolidine-2,4-dione

  • MDL number MFCD09817365
  • Cas Number : 83514-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30712
Availability details for
Quantity
  • Availability
  • Add to Cart