ENA152120957 Enamine

4-methyl-12-oxa-2,3,7-triazatricyclo[7.4.0.0,2,6]trideca-1(9),3,5,7-tetraen-13-one

  • MDL number MFCD09863402
  • Cas Number : 1000932-81-9
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30733
Availability details for
Quantity
  • Availability
  • Add to Cart