ENA152120958 Enamine

4-(cyclopentyloxy)aniline hydrochloride

  • MDL number MFCD09863405
  • Cas Number : 1172521-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30744
Availability details for
Quantity
  • Availability
  • Add to Cart