ENA152120959 Enamine

N-{3-[N-hydroxyethanimidoyl]phenyl}acetamide

  • MDL number MFCD09863406
  • Cas Number : 39569-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30748

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30748
Availability details for
Quantity
  • Availability
  • Add to Cart