ENA152120963 Enamine

N-{4-[1-(hydroxyimino)ethyl]phenyl}cyclopropanecarboxamide

  • Cas Number : 1000932-84-2
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30761
Availability details for
Quantity
  • Availability
  • Add to Cart