ENA152120964 Enamine

N-(2-chloroacetyl)-1-methyl-4-(morpholine-4-sulfonyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD09863411
  • Cas Number : 1000932-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30762

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30762
Availability details for
Quantity
  • Availability
  • Add to Cart