ENA152120970 Enamine

ethyl 5-cyano-1-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylate

  • MDL number MFCD09863417
  • Cas Number : 90878-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30777
Availability details for
Quantity
  • Availability
  • Add to Cart