ENA152120988 Enamine

3-methylpiperidine-1-sulfonyl chloride

  • MDL number MFCD08442451
  • Cas Number : 627887-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29819

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29819
Availability details for
Quantity
  • Availability
  • Add to Cart