ENA152120990 Enamine

N-(3-amino-2-methylphenyl)acetamide

  • MDL number MFCD09037783
  • Cas Number : 65999-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-145
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29821

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29821
Availability details for
Quantity
  • Availability
  • Add to Cart