ENA152120991 Enamine

1-(4-tert-butylphenyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD08444803
  • Cas Number : 923249-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29822
Availability details for
Quantity
  • Availability
  • Add to Cart