ENA152121000 Enamine

1-benzyl-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD00463110
  • Cas Number : 1134-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-67
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29854
Availability details for
Quantity
  • Availability
  • Add to Cart