ENA152121013 Enamine

3-(3-chloropropanoyl)-1-methylurea

  • MDL number MFCD09863313
  • Cas Number : 82277-88-1
  • Molecular Weight : 164.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-29872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29872
Availability details for
Quantity
  • Availability
  • Add to Cart