ENA152121027 Enamine

3-(2,3-dihydro-1,4-benzodioxine-6-sulfonyl)propanoic acid

  • MDL number MFCD07364109
  • Cas Number : 881044-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22526
Availability details for
Quantity
  • Availability
  • Add to Cart