ENA152121030 Enamine

N-(2-chloroacetyl)-1H-pyrrole-2-carboxamide

  • MDL number MFCD09863322
  • Cas Number : 1000931-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29897
Availability details for
Quantity
  • Availability
  • Add to Cart