ENA152121037 Enamine

benzyl(methyl)sulfamoyl chloride

  • MDL number MFCD08443200
  • Cas Number : 36959-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29904

Safety Information

msds https://www.enaminestore.com/catalog/EN300-29904
Availability details for
Quantity
  • Availability
  • Add to Cart