ENA152121046 Enamine

4-(methylsulfamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD09813526
  • Cas Number : 247186-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-105
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-29913

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P405,P501
hcodes C,Xn
MSDS https://www.enaminestore.com/catalog/EN300-29913
pictograms GHS05,GHS07
hstatements H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart