ENA152121050 Enamine

2-chloro-N-(2-acetamidoethyl)acetamide

  • MDL number MFCD09863262
  • Cas Number : 86240-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23129
Availability details for
Quantity
  • Availability
  • Add to Cart