ENA152121054 Enamine

4-cyano-N,N-diethylbenzene-1-sulfonamide

  • MDL number MFCD00510134
  • Cas Number : 33402-63-0
  • Molecular Weight : 238.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24420
Availability details for
Quantity
  • Availability
  • Add to Cart