ENA152121062 Enamine

1-(thiophene-2-carbonyl)piperidine-3-carboxylic acid

  • MDL number MFCD03128018
  • Cas Number : 436093-08-2
  • Molecular Weight : 239.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-29946
Availability details for
Quantity
  • Availability
  • Add to Cart