ENA152121077 Enamine

5-chloro-2-(1H-tetrazol-1-yl)aniline

  • MDL number MFCD03419538
  • Cas Number : 926238-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30393

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30393
Availability details for
Quantity
  • Availability
  • Add to Cart