ENA152121093 Enamine

3-(4-methoxyphenyl)-1-(3-methylphenyl)-1H-pyrazol-5-amine

  • MDL number MFCD08444764
  • Cas Number : 956192-11-3
  • Molecular Weight : 279.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-30414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30414
Availability details for
Quantity
  • Availability
  • Add to Cart