ENA152121094 Enamine

1-(2-methylphenyl)-3-phenyl-1H-pyrazol-5-amine

  • MDL number MFCD00587589
  • Cas Number : 956256-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30415
Availability details for
Quantity
  • Availability
  • Add to Cart