ENA152121101 Enamine

2-(1,3-benzothiazol-2-yl)quinoline-4-carboxylic acid

  • MDL number MFCD02692276
  • Cas Number : 64873-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27832

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27832
Availability details for
Quantity
  • Availability
  • Add to Cart