ENA152121104 Enamine

ethyl [1,2,4]triazolo[1,5-a]pyrimidine-7-carboxylate

  • MDL number MFCD09396707
  • Cas Number : 941203-64-1
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28238
Availability details for
Quantity
  • Availability
  • Add to Cart