ENA152121107 Enamine

(2-morpholin-4-ylphenyl)methanol

  • MDL number MFCD03086181
  • Cas Number : 465514-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30434

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30434
Availability details for
Quantity
  • Availability
  • Add to Cart